480100
Neomycin Sulfate, γ-Irradiated, Tissue Culture Grade - CAS 1405-10-3 - Calbiochem
Sterilized by γ-irradiation. An aminoglycoside antibiotic that inhibits translation by binding to the 30S ribosomal subunit. Potency (anhydrous): ≥600 µg/mg.
Synonym(s):
Neomycin Sulfate, γ-Irradiated, Tissue Culture Grade - CAS 1405-10-3 - Calbiochem
About This Item
Recommended Products
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
protect from light
color
off-white to yellow
solubility
water: 10 mg/mL
shipped in
ambient
storage temp.
−20°C
InChI
1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22?,23+;/m1./s1
InChI key
OIXVKQDWLFHVGR-GQTDVWSESA-N
General description
Biochem/physiol Actions
Translation by binding to the small subunit of prokaryotic ribosomes. Potency (anhydrous): ≥600 µg/mg.
Warning
Reconstitution
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service