Skip to Content
Merck
All Photos(1)

Documents

480001

Millipore

NDSB-195

A non-detergent sulfobetaine. Reported to prevent protein aggregation and facilitate the renaturation of chemically and thermally denatured proteins.

Synonym(s):

NDSB-195, Dimethylethylammonium Propane Sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO3S
CAS Number:
Molecular Weight:
195.28
UNSPSC Code:
12611902
NACRES:
NA.21

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

General description

A non-detergent sulfobetaine. Reported to prevent protein aggregation and facilitate the renaturation of chemically and thermally denatured proteins. Zwitterionic over a wide pH range. Does not absorb significantly in the near-UV range. Easily removed by dialysis.

Application


  • Protein stabilizer, NDSB-195, enhances the dynamics of the β4 -α2 loop of ubiquitin.: This research explores how NDSB-195 stabilizes proteins by enhancing the dynamics of the β4-α2 loop of ubiquitin, providing insights into its mechanism of action as a protein stabilizer (Wang et al., 2016).

  • Suppression of problematic compound oligomerization by cosolubilization of nondetergent sulfobetaines.: The study demonstrates the effectiveness of NDSB-195 in suppressing the oligomerization of problematic compounds, showcasing its utility in biochemical and pharmaceutical applications (Mizukoshi et al., 2015).

  • Interaction of anti-aggregation agent dimethylethylammonium propane sulfonate with acidic fibroblast growth factor.: This article discusses the interaction of NDSB-195 with acidic fibroblast growth factor, emphasizing its role in preventing protein aggregation during biochemical processes (Xiang et al., 2008).

  • Preparation, crystallization and preliminary X-ray diffraction analysis of the DNA-binding domain of the Ets transcription factor in complex with target DNA.: The study uses NDSB-195 in the preparation and crystallization process, enhancing the stability and quality of the DNA-binding domain crystals for X-ray diffraction analysis (Suwa et al., 2008).

Warning

Toxicity: Irritant (B)

Other Notes

Goldberg, M.E., et al. 1996. Folding & Design 1, 21.
Vuillard, L., et al. 1995. Anal. Biochem. 230, 290.
Vuillard, L., et al. 1995. Biochem. J. 305, 337.
Vuillard, L., et al. 1994. FEBS Lett.353, 294.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service