Skip to Content
Merck
All Photos(1)

Key Documents

474426

Sigma-Aldrich

4-Methylumbelliferyl-α-D-glucopyranoside

≥99% (HPLC), solid, α-glucosidase substrate, Calbiochem®

Synonym(s):

4-Methylumbelliferyl-α-D-glucopyranoside, 4-MU-α-D-Glc

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€114.00

€114.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
€114.00

About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€114.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Product Name

4-Methylumbelliferyl-α-D-glucopyranoside, Substrate for fluorogenic assay of α-glucosidase. Has been used in studies of Pompe disease.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

impurities

≤0.01% 4-methylumbelliferone
≤0.02% β-anomer

color

white

solubility

methanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3[o][c](cc(c3cc2)C)=O

InChI

1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15-,16+/m1/s1

InChI key

YUDPTGPSBJVHCN-JZYAIQKZSA-N

General description

Substrate for fluorogenic assay of α-glucosidase. Has been used in studies of Pompe disease.
Substrate for fluorogenic assay of a-glucosidase (excitation maximum: 360 nm; emission maximum: 440 nm). Has been used in studies of Pompe disease.

Warning

Toxicity: Standard Handling (A)

Other Notes

Butterworth, J., and Broadhead, D.M. 1978. Clin. Genet. 6, 504.
Butterworth, J., and Broadhead, D.M. 1977. Clin. Chim. Acta78, 335.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service