Skip to Content
Merck
All Photos(1)

Key Documents

454854

Sigma-Aldrich

MAP Kinase 2, His•Tag®, Human, Recombinant, E. coli

Synonym(s):

ERK2, MAPK2, p42 MAPK, ERK2, MAPK2, p42MAPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥5500 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

Proteases, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human MAP Kinase 2 fused to a His•Tag sequence and expressed in E. coli. Highly active form produced by phosphorylation of the purified MAP Kinase in vitro with MEK1. Suitable for labeling MAP Kinase substrates.
Recombinant, human MAP Kinase 2 fused to a His•Tag sequence and expressed in E. coli. Highly active form produced by phosphorylation of the purified MAP kinase in vitro with MEK1. Suitable for labeling MAP Kinase substrates.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the transfer of 1.0 pmol phosphate to myelin basic protein (MBP) per min at 30°C.

Physical form

In 150 mM NaCl, 50 mM Tris, 1 mM DTT, 50% glycerol, pH 8.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Lew, J. 2003. Biochemistry42, 849.
Johnson, G.L., and Lapadat, R. 2002. Science298, 1911.
Koistinaho, M., and Koistinaho, J. 2002. Glia40, 175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milla Koistinaho et al.
Glia, 40(2), 175-183 (2002-10-16)
Although microglial cells are thought to play a beneficial role in the regeneration and plasticity of the central nervous system (CNS), recent studies have indicated that at least some molecules released by microglia may be harmful in acute brain insults
John Lew
Biochemistry, 42(4), 849-856 (2003-01-29)
Protein kinases constitute one of the largest enzyme families encoded by the human genome. Owing to their critical role in virtually all aspects of signal transduction, protein kinases have evolved stringent mechanisms for their regulation, which classically falls into two
Gary L Johnson et al.
Science (New York, N.Y.), 298(5600), 1911-1912 (2002-12-10)
Multicellular organisms have three well-characterized subfamilies of mitogen-activated protein kinases (MAPKs) that control a vast array of physiological processes. These enzymes are regulated by a characteristic phosphorelay system in which a series of three protein kinases phosphorylate and activate one
Thomas Lanzicher et al.
Frontiers in physiology, 11, 173-173 (2020-04-08)
Titin is a large filamentous protein that forms a sarcomeric myofilament with a molecular spring region that develops force in stretched sarcomeres. The molecular spring has a complex make-up that includes the N2A element. This element largely consists of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service