Skip to Content
Merck
All Photos(1)

Key Documents

442640-M

Sigma-Aldrich

L3MBTL1 Ligand, UNC669

A nicotinamido-pyrrolidine compound that targets the second MBT (Malignant Brain Tumor) module of "chromatin lock" L3MBTL1.

Synonym(s):

L3MBTL1 Ligand, UNC669, 5-Bromo-N-(4-(pyrrolidinyl)piperidinyl)nicotinamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€133.00

€133.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€133.00

About This Item

Empirical Formula (Hill Notation):
C15H20BrN3O
Molecular Weight:
338.24
UNSPSC Code:
12352200
NACRES:
NA.77

€133.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale orange-yellow

solubility

water: 2.5 mg/mL
DMSO: 50 mg/mL

storage temp.

−20°C

General description

A nicotinamido-pyrrolidine compound that targets the second MBT (Malignant Brain Tumor) module of "chromatin lock" L3MBTL1 (Lethal 3 MBT-Like protein 1; Kd = 5 µM by Isothermal titration calorimetry) that "reads"/binds mono- and dimethyl-lysine modified histones as well as other proteins involved in transcription, such as p53 and Rb, via methylation-dependent recognition. Shown to effectively compete against FAM-labeled, monomethylated Histone H3 (1-15) peptide (H3K9Me1) binding to L3MBTL1 (Kd = 10 µM in a FP displacement assay) and exhibit higher affinity toward L3MBTL1 than L3MBTL3 and L3MBTL4 (IC50 = 6, 35, and 69 µM, respectively, in a luminescent peptide competitive binding assay), while displaying no detectable binding toward four other methyl-lysine-binding proteins, the BMT domain-containing SFMBT1 and MBTD1, the PHD (Plant Homeodomain) finger-containing PHF13 (PHD finger 13), and the chromo domain-containing CBX7 (chromobox 7). As of end of 2012, no published data is available regarding the molecule′s permeability or biological activity in cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Herold, J.M., et al. 2011. J. Med. Chem.54, 2504.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service