Skip to Content
Merck
All Photos(1)

Key Documents

438073-M

Sigma-Aldrich

Nitric acid

ACS reagent, 70%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor pressure

8 mmHg ( 20 °C)

Assay

68.0-70.0% (ACS specification)
70%

ign. residue

≤5 ppm

color

APHA: ≤10

bp

120.5 °C (lit.)

density

1.413 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 ppm
sulfate (SO42-): ≤1 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.2 ppm

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid (HNO3) is a highly corrosive mineral acid. It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them. The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight, Raman spectrum, solubility, morphology and alignment examinations.

Application

Nitric acid was used to increase the number of acid sites by altering the surface of activated carbon.
It may be used:
  • In combination with acetic anhydride and zeolite catalysts for the preparation of p-nitrotoluene with high regioselectivity.
  • In the pretreatment of rice straw for enhanced production of xylose.
  • To minimize the extent of cation ordering in LiNi0.5Mn1.5O4, a transition metal-substituted spinel material, by surface treatment.

Other Notes

SKU-Pack sizes with -P are packaged in poly bottles
Important notice
This product will be phased out towards early 2019. There is a replacement product available : 438073

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilgook Kim et al.
Carbohydrate polymers, 99, 563-567 (2013-11-28)
In this study, nitric acid (HNO₃) was evaluated as an acid catalyst for rice straw pretreatment, and, after neutralization, as a sole nitrogen source for subsequent fermentation. Response surface methodology was used to obtain optimal pretreatment condition with respect to
Chemical burn with nitric acid and xanthoproteic reaction.
Daniel A González-Padilla et al.
The Journal of emergency medicine, 48(4), e101-e102 (2014-12-10)
THE TREATMENT OF ACID AND ALKALI BURNS: AN EXPERIMENTAL STUDY.
E C Davidson
Annals of surgery, 85(4), 481-489 (1927-04-01)
Nitric acid purification of single-walled carbon nanotubes.
Hu H, et al.
The Journal of Physical Chemistry B, 107(50), 13838-13842 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service