Skip to Content
Merck
All Photos(1)

Key Documents

437707

Sigma-Aldrich

Lipase, Chromobacterium viscosum

Synonym(s):

Lipase, Chromobacterium viscosum

Sign Into View Organizational & Contract Pricing

Select a Size

100 KU
€272.00

€272.00


Estimated to ship on12 February 2025Details



Select a Size

Change View
100 KU
€272.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€272.00


Estimated to ship on12 February 2025Details


form

solid

Quality Level

specific activity

≥2500 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

pI 

(1) 3.7, (2) 7.3

shipped in

ambient

storage temp.

−20°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

General description

Native lipase from Chromobacterium viscosum. Catalyzes hydrolysis of triglycerides to glycerol and fatty acids.
Native lipase from Chromobacterium viscosum. Catalyzes the hydrolysis of triglycerides to glycerol and fatty acids. Has a pI of 3.7 and 7.3.
Note: 1 KU = 1000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release 1.0 µmol fatty acids from olive oil per min at 37°C, pH 8.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Reconstitute in 100 mM potassium phosphate buffer, pH 8.0, containing 1 mg/ml BSA.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service