Skip to Content
Merck
All Photos(1)

Key Documents

431784

Millipore

Lectin, PHA-L, Phaseolus vulgaris

PHA-L is a potent lymphocyte mitogen with a narrow range. Requires calcium and manganese ions for activity. It has also been found to be useful as a specific marker in anterograde neuronal tracing. Elution: 100 mM acetic acid.

Synonym(s):

Red Kidney Bean Isolectin L

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€260.00

€260.00


Estimated to ship on16 April 2025Details



Select a Size

Change View
5 MG
€260.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.56

€260.00


Estimated to ship on16 April 2025Details


Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Elution: 100 mM acetic acid.
PHA-L is a potent lymphocyte mitogen with a narrow concentration range. Requires calcium and manganese ions for activity. It has also been found to be useful as a specific marker in anterograde neuronal tracing. Elution: 100 mM acetic acid.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to months at -20°C.
Reconstitute in 1 ml 10 mM sodium phosphate buffer, pH 8.0 to yield a final buffer concentration of 15 mM phosphate, 10 mM NaCl with trace Ca+2.

Analysis Note

homogeneous by SDS-PAGE

Other Notes

Chan, F.L., and Ho, S.M. 1999. Prostate 38, 1.
Matsushita, M. 1999. J. Comp. Neurol. 404, 21.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Questions

  1. Comment faut il reconstituer ce produit 

    1 answer
    1. This product is a lyophilized powder and may be reconstituted in 1 mL of 10 mM sodium phosphate buffer, pH 8.0. This will yield a final buffer concentration of 15 mM phosphate, 10 mM NaCl, with trace Ca+2.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service