Skip to Content
Merck
All Photos(1)

Key Documents

427217-M

Sigma-Aldrich

Lactate Dehydrogenase, Rabbit Muscle

Synonym(s):

Lactate Dehydrogenase, Rabbit Muscle, LDH

Sign Into View Organizational & Contract Pricing

Select a Size

427217-25KU
€188.00

€188.00


Estimated to ship on17 April 2025Details



Select a Size

Change View
427217-25KU
€188.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€188.00


Estimated to ship on17 April 2025Details


description

Merck USA index - 14, 5334

Quality Level

form

lyophilized

specific activity

≥200 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.005% GOT and GPT

solubility

low ionic strength buffer: soluble
water: soluble

storage temp.

−20°C

General description

Native lactate dehydrogenase from rabbit muscle. Catalyzes the reversible conversion of pyruvate to lactate. The overall equilibrium of this reaction favors lactate formation. Has an optimal pH 6.0.
Note: 1 KU = 1000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1.0 µmol pyruvate to L-lactate per min in 100 mM phosphate buffer at 30°C, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cahn, R.C., et al. 1962. Science136, 962.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service