Skip to Content
Merck
All Photos(1)

Key Documents

423500

Sigma-Aldrich

Anti-Klotho Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-KL

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€622.00

€622.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€622.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€622.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Kl(16591)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~100 kDa Klotho protein.
Recognizes the ~100 kDa Klotho protein.
This Anti-Klotho Rabbit pAb is validated for use in ELISA, Immunoblotting for the detection of Klotho.

Immunogen

Mouse
a synthetic peptide corresponding to a 17-amino acid sequence near the C-terminus of murine Klotho

Application

ELISA (1:10,000-1:100,000)

Immunoblotting (1-10 µg/ml, chemiluminescence)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.1% BSA, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Kuro-o, M., et al. 1997. Nature390, 45.
Martin, G.M. and Mian, I.S. 1997. Nature390, 18.
Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Feng et al.
Cancer research, 73(8), 2551-2562 (2013-02-27)
Prostate cancer is the most common visceral malignancy and the second leading cause of cancer deaths in US men. There is broad evidence that fibroblast growth factor (FGF) receptors are important in prostate cancer initiation and progression, but the contribution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service