Skip to Content
Merck
All Photos(1)

Key Documents

422686

Sigma-Aldrich

Kininogen, High Molecular Weight, Single Chain, Human Plasma

Native, single chain, high molecular weight Kininogen from human plasma.

Synonym(s):

High molecular weight kininogen

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€688.00

€688.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€688.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.28

€688.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, single chain, high molecular weight Kininogen from human plasma. Nonenzymatic cofactor of the contact activation system. Links prekallikrein to negatively charged surfaces, thereby allowing activation to kallikrein by surface-bound Factor α-XIIa. Also circulates in plasma in complex with Factor XI and may play a role in the activation of Factor XI to Factor XIa by Factor α-XIIa. Serves as a source of bradykinin, a potent vasoactive peptide important in hypotension studies.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 4 mM sodium acetate buffer, pH 5.3.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Weisel, J.W., et al. 1994. J. Biol. Chem. 269, 10100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rasmus Pihl et al.
Science advances, 7(2) (2021-02-02)
Inter-α-inhibitor heavy chain 4 (ITIH4) is a poorly characterized plasma protein that is proteolytically processed in multiple pathological conditions. However, no biological function of ITIH4 has been identified. Here, we show that ITIH4 is cleaved by several human proteases within

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service