Skip to Content
Merck
All Photos(1)

Key Documents

422426

Sigma-Aldrich

KGF/FGF-7, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€483.00

€483.00


Estimated to ship on11 April 2025Details



Select a Size

Change View
25 μG
€483.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€483.00


Estimated to ship on11 April 2025Details


Assay

≥90% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse keratinocyte growth factor (KGF) consisting of amino acids 32-194 and expressed in E. coli. KGF plays an essential role in development, tumorigenesis, angiogenesis, morphogenesis, and wound healing. Mature mouse KGF amino acid sequence is 96% identical with rat, bovine, ovine, equine, and human forms and is active across species.

Biochem/physiol Actions

ED₅₀ = 10-50 ng/ml as measured in a cell proliferation assay using 4MBr-5 monkey epithelial cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered MOPS, Na₂SO₄, EDTA, DTT BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 250 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Belleudi, F., et al. 2007. Traffic8, 1854.
Finch, P.W. and J.S. Rubin. 2006. J. Natl. Cancer Inst.98, 812.
de Georgi, V., et al. 2007. Dermatol. Clin.25, 477.
Niu, J., et al. 2007. J. Biol. Chem.282, 6001.
Werner, S., et al. 2007. J. Invest. Dermatol.127, 998.
Cardinali, G., et al. 2005. J. Invest. Dermatol.125, 1190.
Geer, D.J., et al. 2005. Am. J. Pathol.167, 1575.
Hsu, YR., et al. 1999. Biochemistry38, 2523.
Qiao, J., et al. 1999. Development126, 547.
Werner, S., 1998. Cytokine Growth Factor Rev.9, 153.
Guo, L., et al. 1996. Genes Dev.10, 165.
Mason, I.J., et al. 1994. Mech. Dev.45, 15.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service