Skip to Content
Merck
All Photos(1)

Key Documents

420201

Sigma-Aldrich

JMJD2 Inhibitor, 5-carboxy-8HQ

The JMJD2 Inhibitor, 5-carboxy-8HQ, also referenced under CAS 5852-78-8, controls the biological activity of JMJD2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JMJD2 Inhibitor, 5-carboxy-8HQ, JHDM Inhibitor I, Histone Lysine Demethylase Inhibitor II, 5-carboxy-8HQ, IOX1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H7NO3/c12-8-4-3-7(10(13)14)6-2-1-5-11-9(6)8/h1-5,12H,(H,13,14)

InChI key

JGRPKOGHYBAVMW-UHFFFAOYSA-N

General description

A cell-permeable, 5-carboxy-8-hydroxyquinoline that acts as a 2-oxoglutarate competitive inhibitor of JMJD (IC50 = 200 nM against JMJD2E in a FDH coupled assay). It demonstrates higher selectivity than 2.4-PDCA against other 2-OG oxygenases (IC50 = 2.4 µM, 1.7 µM, 20.5 µM, and 14.3 µM for JMJD2E, JMJD2A, FIH, and PHD2, respectively, in a MALDI-TOF MS assay). Additionally, it is shown to inhibit H3K9me3 demethylation (IC50 = 86.5 µM) in JMJD2A-transfected HeLa cells in a cellular demethylase assay, dose-dependently.
A cell-permeable, 5-carboxy-8-hydroxyquinoline that acts as a 2-oxoglutarate competitive inhibitor of JMJD (IC50 = 200 nM against JMJD2E in a FDH coupled assay). It demonstrates higher selectivity than 2.4-PDCA against other 2-OG oxygenases (IC50 = 2.4 µM, 1.7 µM, 20.5 µM, and 14.3 µM for JMJD2E, JMJD2A, FIH, and PHD2, respectively, in a MALDI-TOF MS assay). Additionally, it is shown to inhibit H3K9me3 demethylation (IC50 = 86.5 µM) in JMJD2A-transfected HeLa cells in a cellular demethylase assay, dose-dependently.




This probe is supplied in conjunction with the Structural Genomics Consortium (SGC). For further characterization details, please visit the IOX1 probe summary on the SGC website.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstiution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hamada, S., et al. 2010. J. Med. Chem.53, 5629.
King, D.N.F., et al. 2010. PLoS One5, e15535.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Hamada et al.
Journal of medicinal chemistry, 53(15), 5629-5638 (2010-08-06)
Selective inhibitors of Jumonji domain-containing protein (JMJD) histone demethylases are candidate anticancer agents as well as potential tools for elucidating the biological functions of JMJDs. On the basis of the crystal structure of JMJD2A and a homology model of JMJD2C
Oliver N F King et al.
PloS one, 5(11), e15535-e15535 (2010-12-03)
Small molecule modulators of epigenetic processes are currently sought as basic probes for biochemical mechanisms, and as starting points for development of therapeutic agents. N(ε)-Methylation of lysine residues on histone tails is one of a number of post-translational modifications that
Ignacio Campillo-Marcos et al.
Frontiers in cell and developmental biology, 9, 715126-715126 (2021-09-21)
Chromatin is dynamically remodeled to adapt to all DNA-related processes, including DNA damage responses (DDR). This adaptation requires DNA and histone epigenetic modifications, which are mediated by several types of enzymes; among them are lysine methyltransferases (KMTs). KMT inhibitors, chaetocin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service