Skip to Content
Merck
All Photos(1)

Key Documents

420123-M

Millipore

JNK Inhibitor II, Negative Control

The JNK Inhibitor II, Negative Control, also referenced under CAS 54642-23-8, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor II, Negative Control, N¹-Methyl-1,9-pyrazoloanthrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

General description

A useful negative control for JNK Inhibitor II (SP600125, Cat. No. 420119). Inhibits JNK2 and JNK3 only at much higher concentrations (IC50 = 18 µM and 24 µM, respectively) compared to SP600125 (IC50 = 40 nM and 90 nM, respectively).
A useful negative control for JNK Inhibitor II (SP600125, Cat. No. 420119). Inhibits JNK2 and JNK3 only at much higher concentrations (IC50 = 18 µM and 24 µM, respectively) compared to SP600125 (IC50 = 40 nM and 90 nM, respectively).

Biochem/physiol Actions

Primary Target
A useful negative control for JNK Inhibitor II (SP600125, Cat. No. 420119)
Target IC50: 18 µM and 24 µM against JNK2 and JNK3, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable up to 3 months at -20°C.

Other Notes

Shin, M., and Boyd, Y.D. 2002. Biochim. Biophys. Acta1589, 311.
Bennett, B.L., et al. 2001. Proc. Natl. Acad. Sci. USA98, 13681.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service