Skip to Content
Merck
All Photos(1)

Key Documents

411520

Sigma-Aldrich

Goat Anti-Human IgE, ε-Chain Specific

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€182.00

€182.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€182.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€182.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

biological source

goat

Quality Level

antibody form

diluted serum

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Goat polyclonal antibody supplied as sterile-filtered, defibrinated, delipidized, and adsorbed serum. Recognizes human IgE ε-chain.
This Goat Anti-Human IgE, ε-Chain Specific is validated for use in Immunoelectrophoresis for the detection of Human IgE, ε-Chain Specific.

Immunogen

Human

Application

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted in 500 mM NaCl, 50 mM Tris-HCl, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for human IgE ε-chain as demonstrated by a single arc when tested against IgE-elevated serum. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sievers et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 47(8), 1057-1068 (2017-03-28)
Microarray-based component-resolved diagnostics (CRD) has become an accepted tool to detect allergen-specific IgE sensitization towards hundreds of allergens in parallel from one drop of serum. Nevertheless, specificity and sensitivity as well as a simultaneous detection of allergen-specific IgG4 , as
Henry R Maun et al.
Cell, 179(2), 417-431 (2019-10-05)
Severe asthma patients with low type 2 inflammation derive less clinical benefit from therapies targeting type 2 cytokines and represent an unmet need. We show that mast cell tryptase is elevated in severe asthma patients independent of type 2 biomarker

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service