Skip to Content
Merck
All Photos(1)

Key Documents

40-999

Millipore

BioTek® MultiFlo FX Multimode Dispenser

6- to 1536-well microplater, Magnetic washer, Milliplex plate washer

Synonym(s):

6- to 1536-well microplater, Magnetic washer, Milliplex plate washer

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€45,490.00

€45,490.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€45,490.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.51

€45,490.00


Please contact Customer Service for Availability

manufacturer/tradename

Agilent

General description

BioTek® 405 MultiFlo with washer and Peristaltic pump with 96-well and 384-well magnetic wash capabilities

MultiFlo FX is an automated multimode reagent dispenser for 6 to 1536 well microplates offering BioTek’s unique Parallel Dispense technology. Up to four independent reagents can be dispensed in parallel without potential carryover. MultiFlo FX becomes a versatile multimode dispenser with the addition of either the RAD technology for random access dispensing to 6 to 384well plates or a wash module for 6 to 384well plate washing. Fast, intuitive programming and operation are via the color touch screen user interface. A MultiFlo FX configured with either RAD technology or the wash module replaces up to five liquid handlers.

Application

2D and 3D cell culture to concentration normalization assays, ELISA, magnetic bead-based assays for 96 and 384 wells

Features and Benefits

  • Multimode dispensing accommodates many functions, takes the place of multiple instruments, and is compact and simple to use.
  • Automated, gentle media exchange with Automated Media Exchange (AMX) washes spheroids and suspension cells without disturbing 2D or 3D cell structures, maintaining the integrity of your assay.
  • Unique RAD technology enables custom-mapped dispensing to individual wells for many applications, including concentration normalization assays, saving setup time
  • Custom manifolds, including fully customizable dispensing and aspirate tube positioning, fluid flow rate, and wash parameters, offer easy configuration
  • Parallel Dispense technology allows up to four reagents to be dispensed in parallel without carryover, enabling easy, flexible dispense operation
  • Cell-friendly angled dispense tips on dispense manifolds and wash manifolds allow reagent to gently dispense down the well sides, keeping cell layers intact even through multiple-step operations for improved assay performance

Legal Information

BioTek is a registered trademark of BioTek/Fischer

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service