407252
Insulin-Like Growth Factor-I, Mouse, Recombinant, E. coli
Synonym(s):
mIGF-I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Assay
≥98% (SDS-PAGE and HPLC)
Quality Level
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
impurities
≤1 ng/μg Endotoxin (ng/μg IGF-I)
shipped in
wet ice
storage temp.
−20°C
General description
Full-length, recombinant, mouse insulin-like growth factor I (IGF-I) expressed in E. coli. IGF-I regulates proliferation and survival signals through the Akt pathway. Mouse IGF-I shares 94% sequence identity with human IGF-I and exhibits cross-species reactivity. Useful in the study of cell proliferation and survival signals.
Biochem/physiol Actions
ED₅₀: ≤2 ng/ml (corresponding to a specific activity of ≥1 x 10⁶ units/mg) as determined by a cell proliferation assay using FDC-P1 cells. For most in vitro applications IGF-I exerts is biological activity in the concentration range of 1-20 ng/ml.
Warning
Toxicity: Standard Handling (A)
Physical form
Sterile-filtered prior to lyophilization. Contains no additives.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions. Aliquots may be stored at 4°C for up to 1 week without loss of biological activity.
Reconstitute with sterile H₂O to a final concentration of 0.1-1 mg/ml. Further dilute with aqueous buffer before use.
Other Notes
Clemmons, D.R., et al. 2006. Curr. Opin. in Pharmacol. 6, 620.
Bell, G.I., et al. 1986. Nucleic Acids Res.14, 7873.
Bell, G.I., et al. 1986. Nucleic Acids Res.14, 7873.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service