Skip to Content
Merck
All Photos(1)

Key Documents

40-50021

Millipore

xMAP® Sheath Fluid PLUS, 20L, RUO

Synonym(s):

xMAP Sheath Fluid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€92.30

€92.30


Estimated to ship on27 June 2025Details



Select a Size

Change View
1 EACH
€92.30

About This Item

UNSPSC Code:
41116010
NACRES:
NC.41

€92.30


Estimated to ship on27 June 2025Details


form

liquid

packaging

pkg of 20 L

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

storage temp.

room temp

General description

The FLEXMAP 3D®, xMAP® INTELLIFLEX RU0, xMAP® INTELLIFLEX-DRSE, and Luminex 200 Instruments require sheath fluid PLUS to serve as the delivery medium to transport the sample to the instrument′s optics.

Application

For use with FlexMAP 3D®, xMAP® INTELLIFLEX RU0 and INTELLIFLEX-DRSE, and Luminex 200 systems.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
FLEXMAP 3D is a registered trademark of Luminex Corp
Luminex 200 is a trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Need help deciding which multiplex assay fits your research? Before you start multiplexing with MILLIPLEX® multiplex kits, check out these tips for how to decide what the best MILLIPLEX® assay is for your research. Also, learn what materials are required to complete multiplex workflows but not provided with the kits.

The newer xMAP® Sheath Fluid PLUS formulation contains 3 modes of microbial action to eliminate potential environmental microbial resistance. See how the preparations compare in this performance comparison of the previous xMAP® Sheath Fluid to the new xMAP® Sheath Fluid PLUS formulation in MILLIPLEX® multiplex assays.

Get even more out of your samples with this Luminex® technology-based mouse cytokine assay – the MILLIPLEX® Mouse Cytokine Expanded Panel 1.

Questions

  1. What is the replacement part number for the discontinued Luminex Sheath Fluid Part # 40-50015, and what are the differences between the replacement and the discontinued part?

    1 answer
    1. Luminex Sheath Fluid, 40-50015 is replaced with Sheath Fluid Plus (40-50021). The new Sheath Fluid formulation includes a mixture of three preservatives, enhancing transit stability and reducing the risk of future acquired microbial resistance.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service