Skip to Content
Merck
All Photos(1)

Key Documents

401132

Sigma-Aldrich

Goat Anti-Human IgA, α-Chain Specific Alkaline Phosphatase Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€339.00

€339.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€339.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

€339.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Immunoaffinity purified goat polyclonal antibody, conjugated to alkaline phosphatase. Recognizes human IgA, α-chain.
This Goat Anti-Human IgA, α-Chain Specific Alk-Phos Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Human IgA, α-Chain Specific.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM Tris, 1 mM MgCl₂, 0.1 mM ZnCl₂, 1% BSA, pH 7.6.

Other Notes

p-Nitrophenyl phosphate, 1.0 mg/ml in 10% diethanolamine, pH 9.8, 1 mM MgCl2 at 25°C, was used as substrate for testing immunoenzyme reactivity. Monospecific for human immunoglobulin A, α-chain as determined by immunoelectrophoresis against normal human serum. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malick M Gibani et al.
Nature medicine, 25(7), 1082-1088 (2019-07-05)
Salmonella Typhi is a human host-restricted pathogen that is responsible for typhoid fever in approximately 10.9 million people annually1. The typhoid toxin is postulated to have a central role in disease pathogenesis, the establishment of chronic infection and human host
Elizabeth A Clutterbuck et al.
Immunology, 119(3), 328-337 (2006-10-28)
Primary immunization of infants with protein-polysaccharide conjugate vaccines induces antipolysaccharide antibody and is highly effective in preventing invasive disease caused by encapsulated bacteria. However, recent experience from the UK indicates that this immunity is not sustained in the absence of
Hyesun Jang et al.
Heliyon, 9(7), e17958-e17958 (2023-07-24)
A growing number of studies indicate that coronavirus disease 2019 (COVID-19) is associated with inflammatory sequelae, but molecular signatures governing the normal versus pathologic convalescence process have not been well-delineated. Here, we characterized global immune and proteome responses in matched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service