Skip to Content
Merck
All Photos(1)

Key Documents

4005-OP

Millipore

Ethylenediaminetetraacetic acid

OmniPur® Grade, >= 99.5%

Synonym(s):

OmniPur® EDTA, (Ethylenedinitrilo)tetraacetic Acid, Ethylenediaminetetraacetic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€68.90
5 KG
€525.00

€68.90


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€68.90
5 KG
€525.00

About This Item

Linear Formula:
(HOOCCH2)2NCH2CH2N(CH2COOH)2
CAS Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

€68.90


Estimated to ship on09 April 2025Details


Request a Bulk Order

grade

reagent

Quality Level

Assay

≥99.5%

form

crystalline powder (Fine)

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

pH

2.5 (23 °C, 10 g/L)

solubility

water: ~0.5 g/L at 20 °C

density

0.86 g/cm3 at 20 °C

cation traces

Fe: ≤0.005%
Mg: ≤0.0005%
heavy metals (as Pb): ≤0.001%

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

SMILES string

N(CCN(CC(=O)O)CC(=O)O)(CC(=O)O)CC(=O)O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

General description

Molecular Biology Grade.

Ash: ≤0.2%
Assay: ≥99.5%
Color of 0.5M Solution (APHA): ≤30
DNAse: none detected
Heavy metals (as Pb): ≤0.001%
Iron (Fe): ≤0.005%
Magnesium (Mg): ≤0.0005%
Nitrilotriacetic acid: ≤0.1%
pH (5%, water): 2.5-3.5
Protease: none detected
RNAse: none detected
Solubility (5%, 1 N NaOH): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

  • Metabolomic Profiling of an Ultraprocessed Dietary Pattern in a Domiciled Randomized Controlled Crossover Feeding Trial.: The research utilizes EDTA in metabolomic profiling to analyze dietary patterns and their impacts on metabolic health, highlighting the utility of EDTA in biochemical analyses (O′Connor et al., 2023).

  • MIL-101(Fe) based biomass as permeable reactive barrier applied to EK-PRB remediation of antimony contaminated soil.: This paper explores the application of EDTA in environmental biochemistry, particularly in the remediation of antimony-contaminated soils using a novel iron-based biomass barrier (Wang et al., 2023).

  • Proteomic signatures of synergistic interactions in antimicrobials.: EDTA is used to study synergistic effects in antimicrobial treatments through advanced proteomic techniques, underscoring its role in biochemistry and molecular biology research (Zhou et al., 2023).

  • Improved Automated Quantification Algorithm (AQuA) and Its Application to NMR-Based Metabolomics of EDTA-Containing Plasma.: The study focuses on the development of an improved algorithm for quantifying metabolites in EDTA-containing plasma samples, demonstrating the importance of EDTA in biochemical quantification and analysis (Röhnisch et al., 2021).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service