Skip to Content
Merck
All Photos(1)

Key Documents

386698

Sigma-Aldrich

Hydrocortisone

≥99% (HPLC), solid, anti-inflammation agent, Calbiochem®

Synonym(s):

Hydrocortisone, Chromatographic Standard, Cortisol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€143.00

€143.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€143.00

About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€143.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Product Name

Hydrocortisone, Chromatographic Standard, Conforms to USP Reference Standard.

Quality Level

description

Merck USA index - 14, 4787

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@]4(CCC(=O)C=C4CC3)C)[C@H](C2)O)CC1)C)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

General description

Conforms to USP Reference Standard.
Steroid synthesized by the adrenal cortex. Important in the regulation of protein, nucleic acid, and lipid metabolism. Pharmacologically, hydrocortisone is used to suppress the inflammatory response. This preparation conforms to the USP Reference Standard.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Conforms to USP Reference Standard
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 month at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Adrenal gland,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fujio Toki et al.
Regenerative therapy, 14, 222-226 (2020-05-22)
Impaired wound healing in critical limb ischemia (CLI) results from multiple factors that affect many cell types and their behavior. Epidermal keratinocytes and dermal fibroblasts play crucial roles in wound healing. However, it remains unclear whether these cell types irreversibly
Sophie Dekoninck et al.
Cell, 181(3), 604-620 (2020-04-08)
During embryonic and postnatal development, organs and tissues grow steadily to achieve their final size at the end of puberty. However, little is known about the cellular dynamics that mediate postnatal growth. By combining in vivo clonal lineage tracing, proliferation kinetics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service