Skip to Content
Merck
All Photos(1)

Key Documents

382158

Sigma-Aldrich

Anti-Histone H3, Acetylated (1-20) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€421.00

€421.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€421.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€421.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

Tetrahymena sp., human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys9,Lys14)

Gene Information

human ... H3C1(8350)

General description

Anti-Histone H3, Acetylated (1-20), rabbit polyclonal, recognizes the ~17 kDa acetylated histone H3 protein. It is validated for use in Western blotting, immunocytochemistry & immunoprecipitation.
Protein A purified rabbit polyclonal antibody. Recognizes the ~17 kDa acetylated histone H3 protein.
Recognizes the ~17 kDa acetylated histone H3 protein.

Immunogen

Human
a diacetylated synthetic peptide [ARTKQTARAcKSTGGAcKAPRKQL(C)] corresponding to amino acids 1-20 of human histone H3, conjugated to KLH

Application

Immunoblotting (10-500 ng/ml, chemiluminescence)

Immunocytochemistry (5 µg/ml)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM Tris-glycine, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HeLa cells

Other Notes

Braunstein, M., et al. 1996. Mol. Cell. Biol.16, 4349.
Braunstein, M., et al. 1993. Genes Devel.7, 592.
Perry, C.A., et al. 1993. Biochemistry32, 13605.
Lin, R., et al. 1989. J. Cell. Biol.108, 1577.
For immunocytochemistry, HeLa cells were fixed with 95% ethanol/5% acetic acid and treated with 5 mM sodium butyrate, an inhibitor of histone deacetylases, for 24 h. 10-500 ng/ml of antibody detected acetylated histone H3 in acid-extracted proteins from HeLa cells. Using sodium butyrate, an inhibitor of histone deacetylases, enhances detection of acetylated histone H3. Higher concentrations of antibody also detected unacetylated histone H3. Preliminary evidence from an independent library suggests that phosphorylation of Ser10 on histone H3 may disrupt epitope recognition by this antibody. This antibody has also been reported to work for immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingang Yuan et al.
Molecular medicine reports, 14(2), 1139-1145 (2016-06-10)
The aim of the present was to evaluate the effects of DNA methylation and histone acetylation on 2,3,7,8‑tetrachlorodibenzo‑p‑dioxin (TCDD)‑induced cleft palate in fetal mice. Pregnant mice (n=10) were randomly divided into two groups: i) TCDD group, mice were treated with 28 µg/kg
Thomas Beckers et al.
International journal of cancer, 121(5), 1138-1148 (2007-04-25)
Advanced second generation inhibitors of histone deacetylases (HDAC) are currently used in clinical development. This study aimed at comparing the pharmacological properties of selected second generation HDAC inhibitors with the hydroxamate and benzamide head group, namely SAHA, LAQ824/LBH589, CI994, MS275
Haiyang Li et al.
eLife, 7 (2018-10-23)
Oomycete pathogens secrete host cell-entering effector proteins to manipulate host immunity during infection. We previously showed that PsAvh52, an early-induced RxLR effector secreted from the soybean root rot pathogen, Phytophthora sojae, could suppress plant immunity. Here, we found that PsAvh52
Atsushi Kuno et al.
Scientific reports, 8(1), 15555-15555 (2018-10-24)
Autophagy activation improves the phenotype in mdx mice, a Duchenne muscular dystrophy (DMD) model, although the underlying mechanisms are obscure. We previously found that resveratrol, a strong inducer of autophagy, ameliorates the cardiac pathology of mdx mice. Autophagy could eliminate
Matías Alvarez-Saavedra et al.
Nature communications, 5, 4181-4181 (2014-06-21)
Chromatin compaction mediates progenitor to post-mitotic cell transitions and modulates gene expression programs, yet the mechanisms are poorly defined. Snf2h and Snf2l are ATP-dependent chromatin remodelling proteins that assemble, reposition and space nucleosomes, and are robustly expressed in the brain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service