Skip to Content
Merck
All Photos(1)

Key Documents

382148

Sigma-Aldrich

Histone Deacetylase Inhibitor II

The Histone Deacetylase Inhibitor II, also referenced under CAS 174664-65-4, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Deacetylase Inhibitor II, CBHA, m-Carboxycinnamic Acid bis-Hydroxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€250.00

€250.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
5 MG
€250.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2O4
CAS Number:
Molecular Weight:
222.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€250.00


Estimated to ship on05 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(O)C(=O)c1cc(ccc1)\C=C\C(=O)NO

InChI

1S/C10H10N2O4/c13-9(11-15)5-4-7-2-1-3-8(6-7)10(14)12-16/h1-6,15-16H,(H,11,13)(H,12,14)/b5-4+

InChI key

OYKBQNOPCSXWBL-SNAWJCMRSA-N

General description

A cell-permeable second generation hybrid polar agent that acts as a histone deacetylase (HDAC) inhibitor. The HDAC inhibition is believed to arise as a result of the binding of the hydroxamic moiety to the active site zinc. Shown to be a potent inducer of transformed cell growth arrest and terminal differentiation (~4 µM). Induces apoptosis and the CD95/CD95-Ligand expression in human neuroblastoma cells, which can be blocked by Caspase Inhibitor Z-VAD-FMK (Cat. No. 627610).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HDAC
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Coffey, D.C., et al. 2001. Cancer Res.61, 3591.
Coffey, D.C., et al. 2000. Pediatr. Oncol.35, 577.
Marks, P.A., et al. 2000. J. Natl. Cancer Inst.92, 1210.
Glick, R.D., et al. 1999. Cancer Res.59, 4392.
Richon, V.M., et al. 1998. Proc. Natl. Acad. Sci. USA95, 3003.
Richon, V.M., et al. 1996. Proc. Natl. Acad. Sci. USA93, 5705.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service