Skip to Content
Merck
All Photos(1)

Key Documents

382065

Sigma-Aldrich

Bisbenzimide H 33342 Fluorochrome, Trihydrochloride

Cell-permeable, adenine-thymine-specific fluorescent stain.

Synonym(s):

Bisbenzimide H 33342 Fluorochrome, Trihydrochloride, 2ʹ-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5ʹ-bi-1H-benzimidazole, 3HCl, HOE 33342

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€126.00

€126.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
100 MG
€126.00

About This Item

Empirical Formula (Hill Notation):
C27H28N6O · 3HCl
CAS Number:
Molecular Weight:
561.93
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.21

€126.00


Estimated to ship on05 May 2025Details


Quality Level

description

RTECS - DT4200000

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

water: 50 mg/mL

fluorescence

λex 346 nm
λem 460 nm

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1(CCN(CC1)c2cc3nc([nH]c3cc2)c4cc5nc([nH]c5cc4)c6ccc(cc6)OCC)C

InChI

1S/C27H28N6O/c1-3-34-21-8-4-18(5-9-21)26-28-22-10-6-19(16-24(22)30-26)27-29-23-11-7-20(17-25(23)31-27)33-14-12-32(2)13-15-33/h4-11,16-17H,3,12-15H2,1-2H3,(H,28,30)(H,29,31)

InChI key

PRDFBSVERLRRMY-UHFFFAOYSA-N

General description

Cell-permeable adenine-thymidine specific fluorescent stain. Useful for staining DNA, chromosomes or nuclei for fluorescence microscopy and flow cytometry applications. DNA-bound dye has an excitation maximum of 358 nm and an emission maximum of 460 nm. Useful for the simultaneous study of the cell cycle and apoptosis in unfixed cells. Induces cell differentiation. H33342 and the related compound H33258 (Cat. No. 382061) have been used in conjunction with RNA aptamers in the 5′ untranslated region (UTR) to control expression of transfected gene constructs. Exhibits a 10 fold greater cell-permeability than H 33258.
Cell-permeable, adenine-thymine-specific fluorescent stain. Useful for staining DNA, chromosomes, and nuclei for fluorescence microscopy and flow cytometry applications. Useful for the simultaneous study of the cell cycle and apoptosis in unfixed cells. Induces cell differentiation. H 33342 and the related compound H 33258 (Cat. No. 382061) have been used in conjunction with RNA aptamers in the 5′ untranslated region (UTR) to control expression of transfected gene constructs. Exhibits a 10-fold greater cell-permeability than H 33258.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Adenine-thymine-specific fluorescent stain
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions (1 mg/ml) are stable for up to 2 weeks at 4°C. Precipitates in phosphate buffer solutions.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Werstuck, G., and Green, M.R. 1998. Science 282, 296.
Paddenberg, R., et al. 1996. Eur. J. Cell Biol. 71, 105.
Filatov, M.V., et al. 1995. Mutat. Res. 327, 209.
Belloc, F., et al. 1994. Cytometry17, 59. Crissman, H.A. 1990. Methods Cell Biol. 33, 97.
Hinkley, R.E., et al. 1986. Dev. Biol.118, 148.
Sandhu, L.C., et al. 1985. Cytometry6, 191.
Frankfurt, O.S. 1983. Cytometry4, 216.
Lalande, M.E., et al. 1981. Proc. Natl. Acad. Sci. USA78, 363.
Visser, J.W.M. 1980. Flow CytometryIV, 86.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Karl Kerns et al.
International journal of molecular sciences, 21(6) (2020-03-25)
Building on our recent discovery of the zinc signature phenomenon present in boar, bull, and human spermatozoa, we have further characterized the role of zinc ions in the spermatozoa's pathway to fertilization. In boar, the zinc signature differed between the
Erin J Plosa et al.
JCI insight, 5(2) (2019-12-25)
Integrins, the extracellular matrix receptors that facilitate cell adhesion and migration, are necessary for organ morphogenesis; however, their role in maintaining adult tissue homeostasis is poorly understood. To define the functional importance of β1 integrin in adult mouse lung, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service