Skip to Content
Merck
All Photos(1)

Key Documents

382061

Sigma-Aldrich

Bisbenzimide H 33258 Fluorochrome, Trihydrochloride

Membrane-permeable, adenine-thymine-specific fluorescent stain.

Synonym(s):

Bisbenzimide H 33258 Fluorochrome, Trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazoyl]-6-(1-methyl-4-piperazyl)-benzimidazole, 3HCl, HOE 33258

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€108.00
250 MG
€232.00
1 G
€676.00

€108.00


In StockDetails



Select a Size

Change View
100 MG
€108.00
250 MG
€232.00
1 G
€676.00

About This Item

Empirical Formula (Hill Notation):
C25H24N6O · 3HCl
CAS Number:
Molecular Weight:
533.88
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.21

€108.00


In StockDetails


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-green

fluorescence

λex 346 nm
λem 460 nm

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[Cl-].N1(CCN(CC1)c2cc3[nH]c(nc3cc2)c4cc5c(cc4)NC(=C6C=CC(=O)C=C6)N5)C.[H+].[H+].[H+]

InChI

1S/C25H24N6O.3ClH/c1-30-10-12-31(13-11-30)18-5-9-21-23(15-18)29-25(27-21)17-4-8-20-22(14-17)28-24(26-20)16-2-6-19(32)7-3-16;;;/h2-9,14-15,26,28H,10-13H2,1H3,(H,27,29);3*1H

InChI key

OKQAASMPIOSOEX-UHFFFAOYSA-N

General description

Membrane-permeable, adenine-thymine-specific fluorescent stain. Useful for staining DNA, chromosomes, and nuclei. Increases the rate of channel-mediated Ca2+ efflux from junctional cytoplasmic reticulum vesicles. Can also be used to detect mycoplasma contamination in tissue culture. H 33258 and the related compound H 33342 (Cat. No. 382065) have been used in conjunction with RNA aptamers in the 5′ untranslated region (UTR) to control expression of transfected gene constructs.
Membrane-permeant, adenine-thymidine specific fluorescent stain (excitation maximum: 346 nm; emission maximum: 460 nm). Useful for staining DNA, chromosomes, and nuclei. Increases the rate of channel-mediated Ca2+ efflux from junctional cytoplasmic reticulum vesicles. Can also be used to detect mycoplasma contamination in tissue culture. H 33258 and the related compound H33342 (Cat. No. 382065) have been used in conjunction with RNA aptamers in the 5′ untranslated region (UTR) to control expression of transfected gene constructs.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Adenine-thymine-specific fluorescent stain
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Preparation Note

Precipitates in phosphate buffered solutions

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Aqueous stock solutions are stable for up to 2 weeks at 4°C.

Other Notes

Werstuck, G., and Green, M.R. 1998. Science 282, 296.
Battaglio, M., et al. 1994. Biotech. Histochem. 69, 152.
Elstein, K.H., and Zucker, R.M. 1994. Exp. Cell Res. 211, 322.
Tarshis, M., et al. 1994. J. Immunol. Methods168, 245.
Beeler, T.J., and Gable, K. 1993. J. Membr. Biol. 135, 109.
Oberhammer, F., et al. 1993. EMBO J. 12, 3679.
Papadimitriou, E., and Lelkes, P.I. 1993. J. Immunol. Methods162, 41.
Jin, R., and Breslauer, K.J. 1988. Proc. Natl. Acad. Sci. USA85, 8939.
Cesarone, C.F., et al. 1979. Anal. Biochem.100, 188.
Latt, S.A., and Stetten, G. 1976. J. Histochem. Cytochem.24, 24.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service