Skip to Content
Merck
All Photos(2)

Key Documents

371822

Sigma-Aldrich

GPR120 Agonist III

The GPR120 Agonist III controls the biological activity of GPR120. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

GPR120 Agonist III, 3-(4-((4-Fluoro-4ʹ-methyl-(1,1ʹ-biphenyl)-2-yl)methoxy)-phenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21FO3
CAS Number:
Molecular Weight:
364.41
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(O)CCC1=CC=C(OCC2=CC(F)=CC=C2C3=CC=C(C)C=C3)C=C1

General description

A membrane-permeant biphenyl-methoxy-phenylpropanoate compound that acts as an active-site targeting, selective, potent agonist of G-protein coupled receptor 120/GPR120 (EC50 of β-arrestin 2 recruitment = 17 nM and 44 nM for mGPR120 and hGPR120, respectively; EC50 of Ca++ mobilization = 96 nM for hGPR120). Exhibits much reduced potency toward free fatty acid receptor 1/FFA1/GPR40 (EC50 of β-arrestin 2 recruitment = 64.6 µM) and no activity toward short chain FFA receptors FFA2/GPR43 and FFA3/GPR41.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
G-protein coupled receptor 120
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shimpukade, B., et al. 2012. J. Med. Chem.55, 4511.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service