Skip to Content
Merck
All Photos(1)

Key Documents

371820

Sigma-Aldrich

G-Protein βγ Binding Peptide, mSIRK(L9A)

A cell-permeable, single point mutated (Leu⁹ to Ala), N-myristoylated G-protein βγ Binding Peptide (mSIRK).

Synonym(s):

G-Protein βγ Binding Peptide, mSIRK(L9A), mSIRK(L9A), myr-SIRKALNIAGYPDYD-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€157.00

€157.00


Estimated to ship on06 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€157.00

About This Item

Empirical Formula (Hill Notation):
C90H144N20O25
Molecular Weight:
1906.23
UNSPSC Code:
12352202
NACRES:
NA.77

€157.00


Estimated to ship on06 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, N-myristoylated G-Protein Binding Peptide (mSIRK) that contains a single point mutation (Leu9 to Ala). In contrast to mSIRK (Cat. No. 371818), the mutant peptide is unable to enhance ERK1/2 phosphorylation. Useful as a negative control for mSIRK.
A cell-permeable, single point mutated (Leu9 to Ala), N-myristoylated G-protein βγ Binding Peptide (mSIRK). Does not enhance ERK1/2 phosphorylation and serves as a negative control for studies involving mSIRK (Cat. No. 371818).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
G-protein βγ
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-N-Ser-Ile-Arg-Lys-Ala-Leu-Asn-Ile-Ala-Gly-Tyr-Pro-Asp-Tyr-Asp-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze(-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Goubaeva, F., et al. 2003. J. Biol. Chem.278, 19634.
Scott, J.K., et al. 2001. EMBO J.20, 767.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service