Skip to Content
Merck
All Photos(1)

Documents

368975

Millipore

Guanidine Thiocyanate

Powerful protein denaturant. Absorbance (1.7 M, H2O, 280 nm):0.30.

Synonym(s):

Guanidine Thiocyanate, GTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N3 · CHNS
CAS Number:
Molecular Weight:
118.16
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.25

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 25%
ethanol: 5%

shipped in

ambient

storage temp.

−20°C

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

General description

Absorbance (1.7 M, H2O, 280 nm): <0.30.
Powerful protein denaturant that is widely used in purification of proteins and nucleic acids (absorbance: <0.30 at 280 nm, 1.7 M, H2O).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service