Skip to Content
Merck
All Photos(1)

Key Documents

367222

Sigma-Aldrich

Gonadotropin, Pregnant Mare Serum

Synonym(s):

PMSG

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

form

solid

color

white

solubility

H2O: soluble

General description

Biopotency: ≥1000 units/mg, as is.
Native gonadotropin from pregnant mare serum. Complex glycoprotein with combined follicle stimulating hormone and interstitial cell-stimulating hormone action.
Native gonadotropin from pregnant mare serum. Complex glycoprotein with combined follicular stimulating hormone and interstitial cell-stimulating hormone action.
This product should not be allowed to come into contact with ruminating animals or swine.

Application

Gonadotropin, Pregnant Mare Serum, CAS 9002-70-4, is a complex glycoprotein with combined follicle stimulating hormone and interstitial cell-stimulating hormone action.

Reconstitution

Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.

Other Notes

Customers in Europe may require an import licence to purchase this product.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S LaPolt et al.
Endocrinology, 130(3), 1289-1295 (1992-03-01)
The actions of gonadotropins on ovarian differentiation are associated with dynamic changes in gonadotropin receptor content, presumably due to modulation of receptor gene expression. The present studies used a reverse transcription-polymerase chain reaction to obtain a rat FSH receptor cDNA
D Bolamba et al.
Journal of animal science, 70(6), 1916-1922 (1992-06-01)
Two experiments were conducted to determine the effects of pregnant mare's serum gonadotropin (PMSG) on follicular populations and ovulation rates in prepuberal gilts with grape-type (GT) and honeycomb-type (HT) ovaries. The follicular populations were determined at 170 d of age
E F Sato et al.
FEBS letters, 303(2-3), 121-125 (1992-06-01)
To investigate the role of superoxide dismutase (SOD) in the ovulatory process, SOD isozymes and their mRNAs were determined in the ovary of 22-day-old rats. After treatment with pregnant mare serum gonadotropin (PMSG) and human chorionic gonadotropin (hCG), ovarian activity
Lan-Tao Gou et al.
Cell, 180(6), 1212-1227 (2020-03-15)
The paternal genome undergoes a massive exchange of histone with protamine for compaction into sperm during spermiogenesis. Upon fertilization, this process is potently reversed, which is essential for parental genome reprogramming and subsequent activation; however, it remains poorly understood how this
Tuğba Ekiz Yılmaz et al.
Acta histochemica, 122(2), 151470-151470 (2019-12-10)
Cyclophosphamide (CYP) is one of the alkylating chemotherapeutic agents and its adverse effects on folliculogenesis in the ovary are well-known due to the previous scientific research on this topic. Magnesium has various effects in organisms, including catalytic functions on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service