Skip to Content
Merck
All Photos(1)

Key Documents

361563

Sigma-Aldrich

GSK-3 Inhibitor XXII, Compound A

Synonym(s):

GSK-3 Inhibitor XXII, Compound A, 6-Methyl-N-[3-[[3-(1-methylethoxy)propyl]carbamoyl]-1H-pyrazol-4-yl]pyridine-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N5O3
Molecular Weight:
345.40
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

green-gray

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable pyrazolopyridine compound that acts as a potent inhibitor against both GSK-3α and GSK-3β (IC50 = 2.3 and 2 nM, respectively, in the presence of 0.5 µM of ATP) with much reduced or little activity toward a panel of 27 other commonly studied kinases. Shown to selectively inhibit the phosphorylation of cellular tau on GSK-specific serine/threonine residues, but not those targeted by non-GSK kinases, both in primary rat cortical neuron cultures (up to 30 µM) in vitro and in mice (up to 26 mg/kg/day p.o.) in vivo. Chronic oral treatment of compound A is also reported to significantly reduce insoluble tau aggregate formation by up to 70% (26 mg/kg/day p.o.) in brain tissues from JNPL3 mice expressing human 4R0N tau with a FTDP-17 (P301L) mutation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Uno. Y., et al. 2009. Brain Res.1296, 148.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service