Skip to Content
Merck
All Photos(1)

Key Documents

361548

Sigma-Aldrich

GSK-3β Inhibitor VII

The GSK-3β Inhibitor VII, also referenced under CAS 99-73-0, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor VII, α-4-Dibromoacetophenone, TPK I Inhibitor, Tau Protein Kinase I Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€163.00

€163.00


Estimated to ship on28 April 2025Details



Select a Size

Change View
5 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C8H6Br2O
CAS Number:
Molecular Weight:
277.94
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.77

€163.00


Estimated to ship on28 April 2025Details


description

RTECS - AM6950000

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

BrCC(=O)c1ccc(cc1)Br

InChI

1S/C8H6Br2O/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2

InChI key

FKJSFKCZZIXQIP-UHFFFAOYSA-N

General description

A phenyl α-bromomethyl ketone compound that acts as a cell-permeable, and non-ATP competitive inhibitor of GSK-3β (IC50 = 0.5 µM). This reactive alkylating agent is selective towards GSK-3β and does not affect PKA activity even at concentrations as high as 100 µM.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 0.5 µM against GSK-3β

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Conde, S., et al. 2003. J. Med. Chem.46, 4631.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service