Skip to Content
Merck
All Photos(1)

Key Documents

346764-M

Millipore

α-D-Glucose-6-phosphate, Monosodium Salt

Synonym(s):

α-D-Glucose-6-phosphate, Monosodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NaO9P
CAS Number:
Molecular Weight:
282.12
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (enzymatic)

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: 1 mg/mL
water: 5 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)O

InChI

1S/C6H13O9P.Na/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;/h1,3-6,8-11H,2H2,(H2,12,13,14);/q;+1/p-1/t3-,4+,5+,6+;/m0./s1

InChI key

OBHLNVXMRZXIII-BTVCFUMJSA-M

General description

A glycolytic intermediate that also acts as a competitive and reversible inhibitor of hexokinase. Useful in creating an ATP-regenerating system. Causes the release of mitochondrially bound hexokinase in muscle.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 6 weeks at -20°C.

Other Notes

Montero-Lomeli, M., and de Meis, L. 1992. J. Biol. Chem.267, 1829.
Rose, I.A., and Warms, J.V. 1967. J. Biol. Chem.242, 1635.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service