Skip to Content
Merck
All Photos(1)

Key Documents

345811

Sigma-Aldrich

Endo-β-Galactosidase, Bacteroides fragilis, Recombinant, E. coli

Endo-β-Galactosidase, Bacteroides fragilis, Recombinant, E. coli, hydrolyzes internal β-galactosidic linkages of oligosaccharides in poly-N-acetyl-lactosamine structures.

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€463.00

€463.00


Estimated to ship on15 April 2025Details



Select a Size

Change View
50 μL
€463.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

€463.00


Estimated to ship on15 April 2025Details


recombinant

expressed in E. coli

Quality Level

conjugate

(Glucosaminoglycan)

form

liquid

specific activity

≥14 U/mL
≥140 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Recombinant, Bacteroides fragilis endo-β-galactosidase expressed in E. coli. Endo-β-Galactosidase is an enzyme that hydrolyzes internal β-galactosidic linkages of oligosaccharides in poly-N-acetyl-lactosamine structures. This enzyme resembles the Escherichia freundii enzyme due to its specificity towards bovine corneal keratan sulphate, milk oligosaccharides and the glycolipids lacto-N-neotetraosylceramide and lacto-N-tetraosylceramide.
Recombinant, Bacteroides fragilis endo-β-galactosidase expressed in E. coli. Endo-β-Galactosidase is an enzyme that hydrolyzes internal β-galactosidic linkages of oligosaccharides in poly-N-acetyl-lactosamine structures. This enzyme resembles the Escherichia freundii enzyme due to its specificity towards bovine corneal keratan sulphate, milk oligosaccharides and the glycolipids lacto-N-neotetraosylceramide and lacto-N-tetraosylceramide. Optimum pH: 5.8.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will liberate one µmol reducing sugar from bovine corneal keratan sulfate per min at 37°C, pH 5.8.

Physical form

Sterile-filtered in 20 mM Tris-HCl, pH 7.5.

Reconstitution

There is no significant loss of activity if left at ambient temperature for several days. The enzyme is active for at least 5 days under the Recommended Reaction Conditions above.

Other Notes

Murata, T., et al. 2003. Eur. J. Biochem.270, 3709.
Scudder, P., et al. 1986. Eur. J. Biochem.157, 365.
Scudder, P., et al. 1984. J. Biol. Chem.259, 6586.
Scudder, P., et al. 1983. Biochem. J.213, 485.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joana Nogueira-Rodrigues et al.
Developmental cell, 57(4), 440-450 (2022-01-06)
Regeneration of adult mammalian central nervous system (CNS) axons is abortive, resulting in inability to recover function after CNS lesion, including spinal cord injury (SCI). Here, we show that the spiny mouse (Acomys) is an exception to other mammals, being

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service