Skip to Content
Merck
All Photos(1)

Key Documents

345758

Sigma-Aldrich

Anti-Ganglioside GM₁, Asialo Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€389.00

€389.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€389.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Ganglioside GM₁, Asialo, rabbit polyclonal, recognizes ganglioside GM₁ asialo.
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes ganglioside GM1 asialo.
Recognizes ganglioside GM1 asialo.

Immunogen

purified, bovine brain asialo-ganglioside GM₁

Application

ELISA (1:500)

TLC Immunoblotting (1:100, colorimetric)

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not cross-react with other carbohydrate epitopes. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service