Skip to Content
Merck
All Photos(1)

Key Documents

34539-M

Millipore

D(+)Galactosamine, Hydrochloride

A powerful inhibitor of hepatic RNA synthesis. Increases the adherence of polymorphonuclear leukocytes to hepatic endothelial cells and induces superoxide production. Induces hepatic damage, causing macrophage infiltration.

Synonym(s):

D(+)Galactosamine, Hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€79.50

€79.50


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
€79.50

About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · xHCl
CAS Number:
Molecular Weight:
179.17 (free base basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

€79.50


Estimated to ship on08 April 2025Details


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 4334

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 25 mg/mL

storage temp.

10-30°C

SMILES string

Cl.N[C@@H]1[C@H]([C@H]([C@H](OC1O)CO)O)O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4+,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-BMZZJELJSA-N

General description

A powerful inhibitor of hepatic RNA synthesis. Increases the adherence of polymorphonuclear leukocytes to hepatic endothelial cells and induces superoxide production. Induces hepatic damage, causing macrophage infiltration.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Komatsu, Y., et al. 1994. Hepatology 20, 1548.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service