Skip to Content
Merck
All Photos(1)

Key Documents

344036-M

Sigma-Aldrich

cFMS Receptor Tyrosine Kinase Inhibitor

The cFMS Receptor Tyrosine Kinase Inhibitor, also referenced under CAS 870483-87-7, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

cFMS Receptor Tyrosine Kinase Inhibitor, 5-(3-Methoxy-4-((4-methoxybenzyl)oxy)benzyl)-pyrimidine-2,4-diamine, CSF-1 Receptor Inhibitor, GW2580

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4O3
CAS Number:
Molecular Weight:
366.41
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

Nc1nc(c(cn1)Cc2cc(c(cc2)OCc3ccc(cc3)OC)OC)N

InChI

1S/C20H22N4O3/c1-25-16-6-3-13(4-7-16)12-27-17-8-5-14(10-18(17)26-2)9-15-11-23-20(22)24-19(15)21/h3-8,10-11H,9,12H2,1-2H3,(H4,21,22,23,24)

InChI key

MYQAUKPBNJWPIE-UHFFFAOYSA-N

General description

A cell-permeable diaminopyrimidine compound that acts as a potent, selective, and ATP-competitive inhibitor of cFMS kinase activity (IC50 = 30 nM) with minimal inhibition towards a panel of 26 other kinases (IC50 >5 µM). Shown to selectively inhibit cFMS-mediated cellular functions in vitro as well as CSF-1-dependent tumor growth in vivo.
A cell-permeable diaminopyrimidine compound that acts as a potent, selective, and ATP-competitive inhibitor of cFMS kinase activity (IC50 = 30 nM) with minimal inhibition towards a panel of 26 other kinases (IC50 >5 µM). Shown to selectively inhibit cFMS-mediated cellular functions in vitro as well as CSF-1-dependent tumor growth in vivo. A 10 mM (1 mg/273 µl) solution of cFMS Receptor Tyrosine Kinase Inhibitor (Cat. No. 344041) in DMSO is also available.

Biochem/physiol Actions

Primary Target
CFMS
Target IC50: 30 nM against cFMS kinase activity

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Conway, J.G., et al. 2005. Proc. Natl. Acad. Sci. USA102, 16078.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service