Skip to Content
Merck
All Photos(1)

Key Documents

343020

Sigma-Aldrich

Flt-3 Inhibitor

The Flt-3 Inhibitor, also referenced under CAS 301305-73-7, controls the biological activity of Flt-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Flt-3 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H20N2O4S
CAS Number:
Molecular Weight:
360.43
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: <4 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1NC(=O)c3cc(c(cc3)OC)OC)C(=O)N)CCCC2

InChI

1S/C18H20N2O4S/c1-23-12-8-7-10(9-13(12)24-2)17(22)20-18-15(16(19)21)11-5-3-4-6-14(11)25-18/h7-9H,3-6H2,1-2H3,(H2,19,21)(H,20,22)

InChI key

FSPQCTGGIANIJZ-UHFFFAOYSA-N

General description

A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor (IC50 = 42 nM) with little effect against a panel of 22 other kinases (IC50 ≥3 µM). Shown to exhibit antiproliferative properties against the human acute myelogenous leukemia cell line MV4-11 (IC50 = 340 nM) expressing constitutively active Flt-3.

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: no
Target IC50: 42 nM against Flt-3

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Patch, R.J., et al. 2006. Bioorg. Med. Chem. Lett.16, 3282.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service