Skip to Content
Merck
All Photos(1)

Key Documents

341253

Sigma-Aldrich

FAAH Inhibitor III, URB937

Synonym(s):

FAAH Inhibitor III, URB937

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O4
Molecular Weight:
354.40
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

gray-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A blood-brain-barrier impermeable p-hydroxyphenylcarbamate compound that acts as a potent and irreversible inhibitor of FAAH activity (IC50 = 26.8 nM rat brain) with excellent selectivity over monoacylglycerol lipases (IC50 >100 µM). Shown to rapidly block mouse liver FAAH activity over brain (ED50 = 0.2 mg/kg vs. 40 mg/kg, s.c.), elevate anandamide and palmitoylethanolamide levels in peripheral tissues (1 mg/kg, i.p.) and exert antinociceptive action. The CNS-penetrant FAAH Inhibitor II, URB597 is also available (Cat. No. 341249).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Clapper, J.R., et al. 2010. Nat. Neurosci.13, 1265.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service