Skip to Content
Merck
All Photos(1)

Key Documents

33-126

Sigma-Aldrich

PIP5-Kinase HTRF Assay

The PIP5 Kinase HTRF Assay kit provides a method for assaying PI(4)P-5 kinases in a homogenous 1920 well format. Suitable for screening of potential inhibitors of these enzymes.

Sign Into View Organizational & Contract Pricing

Select a Size

1920 WELLS
€9,140.00

€9,140.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
1920 WELLS
€9,140.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

€9,140.00


Estimated to ship on05 May 2025Details


Quality Level

species reactivity

human, mouse

manufacturer/tradename

HTRF®
Upstate®

technique(s)

activity assay: suitable (kinase)

detection method

fluorometric (HTRF)

shipped in

dry ice

Application

Used to detect/quantify: PIP5 Kinase

Components

Reaction Buffer (4X)

DM A Detection Reagent

DM C Detection Reagent

DM E Detection Reagent

Phosphatidylinositol 4-phosphate (1mM)

Phosphatidylserine (1mM)

Stop Reagent D

Legal Information

HTRF is a registered trademark of Cis Bio International
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service