Skip to Content
Merck
All Photos(1)

Key Documents

324714

Sigma-Aldrich

Endoproteinase Lys-C, Lysobacter enzymogenes

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

form

lyophilized

Quality Level

solubility

H2O: soluble

General description

Native endoproteinase Lys-C from Lysobacter enzymogenes. Serine protease that specifically hydrolyzes amide or ester bonds at the carboxylic side of lysine in peptides and proteins. Useful for sequence and structural analysis. Inhibited by aprotinin, DFP, leupeptin, and TLCK. Has an optimal pH of 8.5-8.8.

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 μmol of Tos-Gly-Pro-Lys-pNA per min at 25°C, pH 7.7.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 2 days at 4°C or for up to 1 month at -20°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Jekel et al.
Analytical biochemistry, 134(2), 347-354 (1983-10-15)
Endoproteinase Lys-C from Lysobacter enzymogenes, which is commercially available, proved to be useful in the determination of primary structures of proteins. The enzyme preferentially cleaves at the carboxyl side of lysine residues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service