Skip to Content
Merck
All Photos(1)

Key Documents

317639

Sigma-Aldrich

Dipeptidylpeptidase IV, His•Tag®, Human, Recombinant, S. frugiperda

DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, and cell-matrix interactions. Inhibition of DPPIV has been reported to be an effective treatment for type II diabetes.

Sign Into View Organizational & Contract Pricing

Select a Size

5 MI.U.
€658.00

€658.00


Estimated to ship on07 May 2025Details



Select a Size

Change View
5 MI.U.
€658.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

€658.00


Estimated to ship on07 May 2025Details


Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥8 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, and cell-matrix interactions. Inhibition of DPPIV has been reported to be an effective treatment for type II diabetes.
Note: 1 mU = 1 milliunit.
Recombinant, human DPP IV fused at the N-terminus to a His•Tag sequence and expressed in S. frugiperda insect cells. A serine exopeptidase dimer composed of two identical subunits of 110-130 kDa. DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, and cell-matrix interactions. Inhibition of DPPIV has been reported to be an effective treatment for type II diabetes. Note: 1 mU = 1 milliunit.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmole H-Gly-Pro-AMC per min at 37°C, pH 8.0.

Physical form

In 20 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% NaN₃, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Pospisilik, JA, et al. 2003. Diabetes52, 741.
Dobers, J., et al. 2002. Protein Expr. Purif.25, 527.
Marguet, D., et al. 2000. Proc. Natl. Acad. Sci.97, 6874.
Misumi, Y., et al. 1992. Biochim. Biophys. Acta1131, 333.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service