Skip to Content
Merck
All Photos(1)

Key Documents

263458

Millipore

Detergent Variety Pack

Five detergents with widely differing properties suitable for solubilization of membrane-bound proteins.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

shipped in

ambient

storage temp.

10-30°C

General description

A membrane protein is considered solubilized if it is present in the supernatant after one hour centrifugation of lysate or a homogenate at 100,000 x g. In most cases, it is also important that the biological activity of the protein be preserved in the supernatant after solubilization by a detergent. Hence, the appropriate detergent is that which yields the maximum amount of biological active protein in the supernatant. Given a large number of detergents available today, choosing an appropriate detergent can be a difficult process.
Five detergents with widely differing properties suitable for solubilization of membrane-bound proteins.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service