Skip to Content
Merck
All Photos(1)

Documents

238806

Sigma-Aldrich

Cdk2/9 Inhibitor

The Cdk2/9 Inhibitor, also referenced under CAS 507487-89-0, controls the biological activity of Cdk2/9. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk2/9 Inhibitor, (4-(2-Amino-4-methylthiazol-5-yl)pyrimidin-2-yl)-(3-nitrophenyl)amine, Cdk7 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N6O2S
CAS Number:
Molecular Weight:
328.35
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H12N6O2S/c1-8-12(23-13(15)17-8)11-5-6-16-14(19-11)18-9-3-2-4-10(7-9)20(21)22/h2-7H,1H3,(H2,15,17)(H,16,18,19)

InChI key

DYTKVFHLKPDNRW-UHFFFAOYSA-N

General description

A cell-permeable aminopyrimidinyl compound that acts as a potent and ATP-competitive inhibitor of Cdk2/E and Cdk9/T1 (Ki = 2 nM and 4 nM, respectively). It also inhibits GSK-3β, Cdk4/D1, Cdk7/H, Cdk1/B, and Abl at higher concentrations (Ki = 20, 53, 70, 80, and 160 nM, respectively), but exhibits little activity towards 9 other kinases (Ki >1.0 µM). Shown to inhibit phosphorylation of cellular Cdk substrates, pRb and RNA polymerase II, and Cdk2-mediated S-phase transition.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk2/E and Cdk9/T1
Product competes with ATP.
Reversible: no
Target Ki: 2 nM and 4 nM against Cdk2/E and Cdk9/T1, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kontopidis, G., et al. 2006. Chem. Biol.13, 201.
Wang, S., et al. 2004. J. Med. Chem.47, 1662.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service