Skip to Content
Merck
All Photos(1)

Key Documents

236608-M

Sigma-Aldrich

C-Reactive Protein, Human, Recombinant, E. coli

C-Reactive Protein, Human, Recombinant, E. coli, CAS 9007-41-4, is suitable for immunological studies.

Synonym(s):

C-Reactive Protein, Human, Recombinant, E. coli, rhCRP

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€421.00

€421.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€421.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€421.00


Please contact Customer Service for Availability

recombinant

expressed in E. coli

Quality Level

description

Merck USA index - 14, 2603

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

solubility

aqueous buffer: soluble

storage temp.

2-8°C

General description

M.W. 23,000 (monomer); 115,000 (pentamer).
Recombinant, human C-reactive protein expressed in E. coli. CRP is a major acute-phase plasma protein that is dramatically elevated in patients with acute-phase conditions. Composed of five identical noncovalently linked subunits. Exhibits Ca2+-dependent binding to a variety of substrates. Important in diagnosis of rheumatoid arthritis. Suitable for immunological studies. In purified states and in plasma the monomeric form (M.W. ~21 kDa) aggregates as a cyclic pentamer. Intended for use as a calibration standard for assays of CRP, for structural and functional studies, and as an antigen for production of antisera.Calibrated by electroimmunoassay against WHO reference standard or equivalent.

Warning

Toxicity: Standard Handling (A)

Physical form

In 140 mM NaCl, 20 mM Tris-HCl, 2 mM CaCl₂, 0.05% NaN₃, pH 7.5.

Other Notes

Paffen, E., et al. 2004. Arterioscler. Thromb. Vasc. Biol.24, 975.
Ballou, S.P., and Kushner, I. 1992. Adv. Intern. Med. 37, 313.
Stults, N.L., et al. 1987. Anal. Biochem. 161, 567.
Woo, P., et al. 1985. J. Biol. Chem. 260, 13384.
de Beer, F.C., and Pepys, M.B. 1982. J. Immun. Methods 50, 17.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the concentration of CRP in the product?

    1 answer
    1. Protein concentration is 0.5 mg/mL - 2.0 mg/mL. Batch-specific concentration can be found in the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service