Skip to Content
Merck
All Photos(2)

Key Documents

233624

Sigma-Aldrich

Cryptocrome Activator, KL001

The Cryptocrome Activator, KL001 modulates the biological activity of Cryptocrome. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

Cryptocrome Activator, KL001, N-(3-(9H-Carbazol-9-yl)-2-hydroxypropyl)-N-(furan-2-ylmethyl)methanesulfonamide, CRY Activator, KL001

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€242.00

€242.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€242.00

About This Item

Empirical Formula (Hill Notation):
C21H22N2O4S
CAS Number:
Molecular Weight:
398.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€242.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(CN(CC1=CC=CO1)S(=O)(C)=O)CN2C3=C(C=CC=C3)C4=C2C=CC=C4

InChI

1S/C21H22N2O4S/c1-28(25,26)22(15-17-7-6-12-27-17)13-16(24)14-23-20-10-4-2-8-18(20)19-9-3-5-11-21(19)23/h2-12,16,24H,13-15H2,1H3

InChI key

OQAFDLPAPSSOHY-UHFFFAOYSA-N

General description

A cell-permeable carbazole compound that competes against FAD (flavin adenine dinucleotide) for cryptocrome (CRY) 1 & 2 binding, displaying little affinity toward other clock proteins (PER1/2, CLOCK, or BMAL1) or inhibitory activity against casein kinase (CKIα, CKIδ, and CK2) activity even at concentrations as high as 100 µM. Shown to selectively stabilize cellular CRY1/2 protein, but not mRNA, by preventing FBXL3-mediated CRY1/2 ubiquitination and proteasomal degradation, effectively lengthening the period and reducing the amplitude of the circadian transcription rhythms of Bmal1 and Per2 promoter-mediated transcription activity (IC50 of intensity reduction = 14 and 0.82 µM, respectively, in U2OS reporter assay). KL001 is also reported to block glucagon-induced Pck1 and G6pc mRNA upregulation as well as glucose production in primary murine hepatocyte cultures (IC50<8 µM). Longdaysin (Cat. No. 438075), in comparison, exerts its circadian perturbation activity via casein kinase inhibition.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cryptocrome protein
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hirota, T., et al. 2012. Science337, 1094.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How can you tell if you've been Xposed to this agent

    1 answer
    1. Please review the product Safety Data Sheet; Section 8 lists exposure controls as well as personal protection recommendations: https://www.sigmaaldrich.com/sds/mm/233624.
      For additional questions please contact your site EHS group. If experiencing symptoms contact a medical professional.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service