Skip to Content
Merck
All Photos(1)

Key Documents

233156

Millipore

Cleland′s Reagent, Molecular Biology Grade

A protective agent for SH groups.

Synonym(s):

Cleland’s Reagent, Molecular Biology Grade, 1,4-Dithiothreitol, DTT

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€77.70
10 G
€371.00

€77.70


Estimated to ship on04 February 2026Details


Request a Bulk Order

Select a Size

Change View
1 G
€77.70
10 G
€371.00

About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12161504
NACRES:
NA.25

€77.70


Estimated to ship on04 February 2026Details


Request a Bulk Order

grade

Molecular Biology

Quality Level

Assay

≥99% (titration)

form

powder
solid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 15 mg/mL

cation traces

heavy metals: ≤0.001%

absorption

≤0.100 at 280 nm at 100 mM (1 cm)
≤0.500 at 260 nm at 100 mM (1 cm)

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

2-8°C

SMILES string

SC[C@H](O)[C@@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

General description

A protective agent for SH groups.
Water-soluble protective reagent for sulfhydryl groups [absorbance (100 mM): ≤0.5 at 260 nm; ≤0.1 at 280 nm]. It is capable of maintaining monothiols completely in the reduced state and of reducing disulfides quantitatively. Since it readily permeates cell membranes, it can protect protein sulfhydryls and restore enzyme activity lost by oxidation.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service