Skip to Content
Merck
All Photos(2)

Key Documents

23-058-M

Sigma-Aldrich

Smurf1, active

Synonym(s):

SMAD ubiquitination regulatory factor 1, SMAD-specific E3 ubiquitin-protein ligase 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€707.00
250 μG
€4,000.00

€707.00


Estimated to ship on24 March 2025Details



Select a Size

Change View
10 μG
€707.00
250 μG
€4,000.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€707.00


Estimated to ship on24 March 2025Details


biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Smurf1 is a member of the Nedd4-like family of E3 ligases. It contains an amino terminal C2 domain and 2 WW domains that are responsible for cellular localization and substrate recognition, and a catalytic carboxyl terminal HECT domain.
Smurf1 was originally identified as a negative regulator of the TGF-β/BMP (bone morphogenetic proteins) signalling pathway by degrading SMAD proteins and TGF-β /BMP receptors. To date, over a dozen substrates have been identified, including Runx2, MEKK2, RhoA, Par6, JunB, Talin head, Prickle1, TRAF family proteins, and STAT1. This suggests a pleiotropic role for Smurf1 in various cellular functions such as bone homeostasis, embryonic development, cell movement and polarity control, and immune response. Aberrant expression of Smurfs has been shown to occur in several types of cancers, including breast, oesophageal, pancreatic and renal cell carcinomas.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using glutathione sepharose.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service