Skip to Content
Merck
All Photos(1)

Key Documents

23-055-M

Sigma-Aldrich

Cardiac Troponin I, 10 µg

Full length, recombinant human Cardiac Troponin I. N-terminal c-Myc, GST-tagged. For use in Enzyme Assays. Functions as a substrate in ubiquitination assays.

Synonym(s):

Troponin I, cardiac muscle, TNNI3, TNNC1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€679.00
250 μG
€3,910.00

€679.00


Estimated to ship on30 April 2025Details



Select a Size

Change View
10 μG
€679.00
250 μG
€3,910.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€679.00


Estimated to ship on30 April 2025Details


biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Troponin is a complex of three regulatory proteins (Troponin I, Troponin T and Troponin C). It is located on the actin filament and is essential for the calcium-mediated regulation of skeletal and cardiac muscle contraction. There are tissue-specific isoforms of Troponin I, T and C and two subtypes (Cardiac Troponin I and T) are released when the heart muscle has been damaged, such as during a heart attack. The more damage there is to the heart, the greater the amount of Troponin I and T there will be in the blood and currently, Cardiac Troponin I and T are used as the preferred biochemical markers for the diagnosis of myocardial infarction. It has also been demonstrated that Cardiac Troponin I is targeted for degradation by MuRF1, an E3 ubiquitin ligase that has been linked to muscle atrophy

Application

Ubiquitination Cascade Comoponent: Substrate

Physical form

Purified using immobilized metal affinity chromatography.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service