Skip to Content
Merck
All Photos(2)

Key Documents

23-044-M

Sigma-Aldrich

VHL Protein Complex, Active, 10 µg

Active complex of five (5) recombinant human enzymes: VHL amino acids 54-end, full length Elongin C, full length Elongin B, full length Cul2, & full length Rbx1. For use in Enzyme Assays. Functions as an E3 ligase in ubiquitination assays.

Synonym(s):

VCB-Cul2 complex, ECV complex, CBC VHL

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€682.00
250 μG
€3,910.00

€682.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€682.00
250 μG
€3,910.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€682.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

solubility

water: soluble

Gene Information

General description

Research area: Cancer

The von-Hippel Lindau tumor suppressor (VHL) complex is a multi-subunit ubiquitin ligase composed of VHL, Elongin B, Elongin C, Cul2 and Rbx1. The VHL protein serves as the substrate recognition component and is linked by Elongin C to a heterodimeric Cul2/Rbx1 module that functions as a potent activator of the ubiquitination of target proteins by an E2 conjugating enzyme. Elongin B interacts with the complex through Elongin C and appears to stabilize the binding of Elongin C to VHL.

Application

Ubiquitination Cascade Comoponent: E3

Biochem/physiol Actions

The primary function of the von-Hippel Lindau tumor suppressor (VHL) complex is to regulate HIF (hypoxia inducible factor) activity by targeting the hydroxylated HIF-1α subunit for ubiquitination and rapid proteasomal degradation during normoxia under normoxic conditions. It therefore plays an important role in the regulation of hypoxia-inducible genes such as the vascular endothelial growth factor (VEGF) and glucose transport-1 (Glut-1). Mutations in VHL are associated with the inherited von Hippel-Lindau (VHL) cancer syndrome and numerous forms of renal cell carcinoma.[1]

Physical form

Purified using glutathione sepharose.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volker H Haase
Current pharmaceutical design, 15(33), 3895-3903 (2009-08-13)
Hypoxia-Inducible Factors (HIFs) are heterodimeric oxygen-sensitive basic helix-loop-helix transcription factors that play central roles in cellular adaptation to low oxygen environments. The von-Hippel Lindau tumor suppressor (pVHL) is the substrate recognition component of an E3 ubiquitin ligase and functions as
Sang Bae Lee et al.
Molecular cell, 79(3), 376-389 (2020-07-09)
Activation of dual-specificity tyrosine-phosphorylation-regulated kinases 1A and 1B (DYRK1A and DYRK1B) requires prolyl hydroxylation by PHD1 prolyl hydroxylase. Prolyl hydroxylation of DYRK1 initiates a cascade of events leading to the release of molecular constraints on von Hippel-Lindau (VHL) ubiquitin ligase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service