Skip to Content
Merck
All Photos(1)

Key Documents

23-043-M

Sigma-Aldrich

Kit Protein, 10 µg

Synonym(s):

CD_antigen=CD117, Mast/stem cell growth factor receptor, Proto-oncogene c-Kit, Tyrosine-protein kinase Kit

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€3,700.00

€3,700.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
€3,700.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€3,700.00


Please contact Customer Service for Availability

biological source

human

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

AXIS Axon Isolation Devices, 150 μm long channels, 10 devices
Kit belongs to the family of type III receptor tyrosine kinases. It mediates the cellular response to stem cell factor and plays a major role in survival, expansion and differentiation of haematopoietic progenitor cells and reproductive stem cells. The activation of Kit is tightly regulated and uncontrolled Kit activity contributes to the growth of diverse human tumours. Gain-of-function mutations have been described in a number of malignancies, including acute myeloid leukaemia, small cell lung cancer, gastrointestinal stromal tumours and testicular cancer. Negative regulation of Kit signalling has been shown to occur mainly through ubiquitin-mediated internalization and degradation of the receptor.

Application

Recombinant human Kit, amino acids 544-end. N-terminal c-Myc, GST-tagged. For use in Enzyme Assays. Functions as a substrate in ubiquitination assays.
Ubiquitination Cascade Comoponent:Substrate

Physical form

Format: Purified using glutathione sepharose.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service